Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDML
Entrez Gene ID
693511
Gene Name
R3H domain containing like
Gene Aliases
-
Chromosome Location
Chr.10: 79559405 - 79575791 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001081974.4 XP_001081974.2 - 1230 147

View Full Product Details