Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD7
Entrez Gene ID
697223
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
EGK_11916
Chromosome Location
Chr.2: 74847275 - 74874240 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028843461.1 XP_028699294.1 - 1078 104

View Full Product Details