Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLT1A
Entrez Gene ID
700305
Gene Name
golgi transport 1A
Gene Aliases
EGK_01763
Chromosome Location
Chr.1: 61704891 - 61721278 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001098173.4 XP_001098173.1 - 536 104

View Full Product Details