Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP3
Entrez Gene ID
713093
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
FAM5C
Chromosome Location
Chr.1: 47371747 - 47769834 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261478.1 NP_001248407.1 - 1026 151
XM_015121080.2 XP_014976566.1 - 1439 151
XM_015121085.2 XP_014976571.1 - 1534 151
XM_015121086.2 XP_014976572.1 - 1172 151
XM_015121094.2 XP_014976580.1 - 942 151
XM_015121101.2 XP_014976587.1 - 1064 151
XM_015121103.2 XP_014976589.1 - 1344 151
GenBank mRNA JU320869.1 - - 1026 151
JV466999.1 - - 1190 151
JV489310.1 - - 1189 151

View Full Product Details