Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL11A
Entrez Gene ID
710394
Gene Name
RAS like family 11 member A
Gene Aliases
EGK_09191
Chromosome Location
Chr.17: 6575194 - 6583135 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194015.2 NP_001180944.1 - 528 116
XM_028837146.1 XP_028692979.1 - 3634 116
GenBank mRNA JU334442.1 - - 331 116
JU470733.1 - - 250 116
JV044256.1 - - 219 116
JV639148.1 - - 788 116

View Full Product Details