Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL9
Entrez Gene ID
695486
Gene Name
ADP ribosylation factor like GTPase 9
Gene Aliases
-
Chromosome Location
Chr.5: 77087346 - 77107029 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015138660.2 XP_014994146.2 - 893 153
GenBank mRNA JU323228.1 - - 164 153

View Full Product Details