Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10G3
Entrez Gene ID
707265
Gene Name
olfactory receptor, family 10, subfamily G, member 3
Gene Aliases
-
Chromosome Location
Chr.7: 83463817 - 83464758 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194096.1 NP_001181025.1 - 891 148

View Full Product Details