Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IFIT1
- Entrez Gene ID
-
695683
- Gene Name
- interferon induced protein with tetratricopeptide repeats 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 89628584 - 89669645 on Build Mmul_10
- Gene Symbol
- LOC114669827
- Entrez Gene ID
-
114669827
- Gene Name
- interferon-induced protein with tetratricopeptide repeats 1B-like
- Gene Aliases
-
IFIT1
- Chromosome Location
-
Chr.9: 89643475 - 89650853 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU334336.1 |
- |
- |
1609 |
110 |
View Full Product Details