Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INHBB
Entrez Gene ID
574343
Gene Name
inhibin subunit beta B
Gene Aliases
-
Chromosome Location
Chr.12: 10750875 - 10756576 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090729.4 XP_001090729.4 - 492 114

View Full Product Details