Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD10
Entrez Gene ID
699597
Gene Name
programmed cell death 10
Gene Aliases
-
Chromosome Location
Chr.2: 72502832 - 72554570 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266410.1 NP_001253339.1 - 312 141
XM_015131604.2 XP_014987090.1 - 305 141
XM_015131606.2 XP_014987092.1 - 329 141
GenBank mRNA JU332745.1 - - 312 141
JU473132.1 - - 312 141
JV046074.1 - - 312 141
JV601896.1 - - 339 141

View Full Product Details