The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047152.1 | NP_001040617.1 | - | 691 | 88 |
XM_015149361.2 | XP_015004847.1 | - | 1368 | 88 | |
XM_015149362.2 | XP_015004848.1 | - | 1364 | 88 | |
XM_015149363.2 | XP_015004849.1 | - | 943 | 88 | |
XM_015149365.2 | XP_015004851.1 | - | 1363 | 88 | |
GenBank mRNA | JV551155.1 | - | - | 493 | 88 |
JV635775.1 | - | - | 795 | 88 |