Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK9
Entrez Gene ID
708270
Gene Name
serine peptidase inhibitor Kazal type 9
Gene Aliases
EGK_17001
Chromosome Location
Chr.6: 145857805 - 145862069 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193441.1 NP_001180370.1 - 133 77

View Full Product Details