Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDF
Entrez Gene ID
100427223
Gene Name
peptide deformylase, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.20: 56581599 - 56583519 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194842.3 NP_001181771.2 - 573 62
GenBank mRNA JU326582.1 - - 159 62
JU471628.1 - - 588 62
JV602396.1 - - 582 62

View Full Product Details