Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMCO3
Entrez Gene ID
699680
Gene Name
single-pass membrane protein with coiled-coil domains 3
Gene Aliases
C11H12orf69, C12orf69
Chromosome Location
Chr.11: 16198292 - 16206319 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190832.1 NP_001177761.1 - 550 92
XM_015151100.2 XP_015006586.1 - 755 92
XM_028828832.1 XP_028684665.1 - 893 92
XM_028828833.1 XP_028684666.1 - 1035 92

View Full Product Details