Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GXYLT2
Entrez Gene ID
693538
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
GLT8D3
Chromosome Location
Chr.2: 128972637 - 129053056 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193691.3 NP_001180620.1 - 602 62
XM_028843068.1 XP_028698901.1 - 760 62
GenBank mRNA JV070198.1 - - 408 62

View Full Product Details