Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOA1
Entrez Gene ID
693437
Gene Name
nitric oxide associated 1
Gene Aliases
C4orf14, C5H4orf14, EGK_15861
Chromosome Location
Chr.5: 76654021 - 76667315 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193688.2 NP_001180617.1 - 1349 98
GenBank mRNA JU323405.1 - - 1520 98
JU477487.1 - - 1541 98
JV663342.1 - - 1575 98

View Full Product Details