Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP11B1
Entrez Gene ID
695793
Gene Name
cytochrome P450, family 11, subfamily B, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.8: 143164731 - 143170343 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193738.3 NP_001180667.2 - 257 74

View Full Product Details