Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC61
Entrez Gene ID
704779
Gene Name
leucine rich repeat containing 61
Gene Aliases
-
Chromosome Location
Chr.3: 176272154 - 176286446 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193828.3 NP_001180757.1 - 292 91
XM_001088845.4 XP_001088845.1 - 535 91
XM_028844911.1 XP_028700744.1 - 316 91
GenBank mRNA JU332137.1 - - 295 91
JU471914.1 - - 502 91
JV555494.1 - - 527 91

View Full Product Details