Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCR4
Entrez Gene ID
707329
Gene Name
C-X-C motif chemokine receptor 4
Gene Aliases
-
Chromosome Location
Chr.12: 21291083 - 21294316 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042645.1 NP_001036110.1 - 46 74
GenBank mRNA AF001928.1 - - 46 74
JU477147.1 - - 115 74
JV636369.1 - - 91 74

View Full Product Details