Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR55
Entrez Gene ID
100862846
Gene Name
WD repeat domain 55
Gene Aliases
EGK_16926
Chromosome Location
Chr.6: 138046100 - 138052488 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256898.2 NP_001243827.2 - 313 87
GenBank mRNA JU326745.1 - - 180 87
JU473793.1 - - 240 87
JU473794.1 - - 240 87
JU473795.1 - - 240 87
JV045452.1 - - 236 87
JV654618.1 - - 296 87

View Full Product Details