Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DEDD
- Entrez Gene ID
-
719875
- Gene Name
- death effector domain containing
- Gene Aliases
-
EGK_01526
- Chromosome Location
-
Chr.1: 89814903 - 89826345 on Build Mmul_10
- Gene Symbol
- NIT1
- Entrez Gene ID
-
720272
- Gene Name
- nitrilase 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 89826131 - 89829246 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU478233.1 |
- |
- |
752 |
73 |
JV048911.1 |
- |
- |
697 |
73 |
View Full Product Details