Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLCD3B
Entrez Gene ID
708210
Gene Name
TLC domain containing 3B
Gene Aliases
FAM57B
Chromosome Location
Chr.20: 27326931 - 27339522 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261178.1 NP_001248107.1 - 657 113
XM_015139856.2 XP_014995342.1 - 989 113
XM_015139858.2 XP_014995344.1 - 465 113
GenBank mRNA JU336630.1 - - 657 113
JV601192.1 - - 786 113

View Full Product Details