Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM11
Entrez Gene ID
694150
Gene Name
tripartite motif containing 11
Gene Aliases
-
Chromosome Location
Chr.1: 85970199 - 85983313 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261480.1 NP_001248409.1 - 662 74
GenBank mRNA JU477353.1 - - 662 74
JV594909.1 - - 311 74

View Full Product Details