Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ8B
Entrez Gene ID
702379
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, EGK_10633
Chromosome Location
Chr.19: 39910633 - 39936815 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265945.1 NP_001252874.1 - 1047 93
XM_015123954.2 XP_014979440.2 - 1378 93
XM_015123956.2 XP_014979442.2 - 1199 93
XM_028838385.1 XP_028694218.1 - 1075 93
XM_028838386.1 XP_028694219.1 - 1492 93
GenBank mRNA JU323098.1 - - 1039 93
JU471714.1 - - 1037 93
JV048291.1 - - 1047 93

View Full Product Details