Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ8B
Entrez Gene ID
702379
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, EGK_10633
Chromosome Location
Chr.19: 39910633 - 39936815 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265945.1 NP_001252874.1 - 117 93
GenBank mRNA JU323098.1 - - 109 93
JU471714.1 - - 107 93
JV048291.1 - - 117 93

View Full Product Details