Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ8B
Entrez Gene ID
702379
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, EGK_10633
Chromosome Location
Chr.19: 39910633 - 39936815 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265945.1 NP_001252874.1 - 726 84
XM_015123954.2 XP_014979440.2 - 1057 84
XM_015123956.2 XP_014979442.2 - 878 84
XM_028838385.1 XP_028694218.1 - 754 84
XM_028838386.1 XP_028694219.1 - 1171 84
GenBank mRNA JU323098.1 - - 718 84
JU471714.1 - - 716 84
JV048291.1 - - 726 84

View Full Product Details