Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN3
Entrez Gene ID
707572
Gene Name
transgelin 3
Gene Aliases
EGK_11281
Chromosome Location
Chr.2: 164165065 - 164180377 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266904.1 NP_001253833.1 - 713 91
XM_015129798.2 XP_014985284.1 - 1052 91
GenBank mRNA JU334168.1 - - 713 91

View Full Product Details