Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIR24-2
Entrez Gene ID
100315495
Gene Name
microRNA mir-24-2
Gene Aliases
mir-24-2, mml-mir-24-2
Chromosome Location
Chr.19: 13435438 - 13435510 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_032089.1 - 31 61

View Full Product Details