The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013107.1 | NP_037239.1 | 2 - 3 | 832 | 122 |
GenBank mRNA | AY184240.1 | - | 2 - 3 | 832 | 122 |