Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt4
Entrez Gene ID
64440
Gene Name
synaptotagmin IV
Gene Aliases
-
Chromosome Location
Chr.18: 24172580 - 24181989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031693.2 NP_113881.1 1 - 2 277 74
GenBank mRNA CB730279.1 - 1 - 2 324 74
CB786178.1 - 1 - 2 267 74
FQ211592.1 - 1 - 2 274 74
L38247.1 - 1 - 2 278 74
U14398.1 - 1 - 2 306 74

View Full Product Details