The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033670.1 | NP_001028842.1 | 2 - 3 | 741 | 65 |
XM_006235265.2 | XP_006235327.1 | 2 - 3 | 1553 | 65 | |
GenBank mRNA | BC094213.1 | - | 2 - 3 | 741 | 65 |
U72350.1 | - | 1 - 2 | 634 | 65 |