Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oprl1
Entrez Gene ID
29256
Gene Name
opiate receptor-like 1
Gene Aliases
KOR-3, KOR3, LC132, MOR-C, OFQR, ORL1, Oprl, XOR1
Chromosome Location
Chr.3: 177225366 - 177231663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031569.3 NP_113757.1 2 - 3 462 92
XM_006235736.2 XP_006235798.1 3 - 4 552 92
XM_006235738.2 XP_006235800.1 2 - 3 310 92
GenBank mRNA AY152731.1 - 1 - 2 221 92
D16438.1 - 2 - 3 273 92
L28144.1 - 2 - 3 393 92
L29419.1 - 1 - 2 356 92
L33916.1 - 2 - 3 327 92
U01913.1 - 2 - 3 402 92
U05239.1 - 2 - 3 404 92
U07871.1 - 1 - 2 252 92

View Full Product Details