Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyth2
Entrez Gene ID
116692
Gene Name
cytohesin 2
Gene Aliases
Arno, Pscd2, Sec7, Sec7B
Chromosome Location
Chr.1: 101796349 - 101803204 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053911.2 NP_446363.1 4 - 5 538 96
XM_006228964.2 XP_006229026.1 4 - 5 556 96
XM_008759304.1 XP_008757526.1 3 - 4 1029 96
GenBank mRNA FQ229744.1 - 3 - 4 259 96
U83896.1 - 4 - 5 542 96

View Full Product Details