The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012805.2 | NP_036937.2 | - | 797 | 75 |
GenBank mRNA | AM295012.1 | - | - | 797 | 75 |
AM295013.1 | - | - | 797 | 75 | |
AM392401.1 | - | - | 797 | 75 | |
L06482.1 | - | - | 964 | 75 |