Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calr
Entrez Gene ID
64202
Gene Name
calreticulin
Gene Aliases
-
Chromosome Location
Chr.19: 25956771 - 25961666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022399.2 NP_071794.1 3 - 4 461 62
GenBank mRNA BC062395.1 - 3 - 4 438 62
BU671467.1 - 3 - 4 461 62
D78308.1 - 3 - 4 413 62
FQ210587.1 - - 466 62
FQ220096.1 - 3 - 4 467 62
X53363.1 - 3 - 4 433 62
X79327.1 - 3 - 4 411 62

View Full Product Details