Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf5l
Entrez Gene ID
307927
Gene Name
TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor
Gene Aliases
-
Chromosome Location
Chr.19: 56781854 - 56801005 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107442.1 NP_001100912.1 2 - 3 322 66
XM_006255812.2 XP_006255874.1 2 - 3 1368 66
XM_008772614.1 XP_008770836.1 2 - 3 188 66
XM_008772615.1 XP_008770837.1 2 - 3 364 66
GenBank mRNA BC160901.1 - 2 - 3 202 66

Gene Symbol
LOC690283
Entrez Gene ID
690283
Gene Name
similar to TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor
Gene Aliases
-
Chromosome Location
Chr.1: 60105788 - 60108103 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758820.1 XP_008757042.1 1 - 1 387 66

View Full Product Details