Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chaf1a
Entrez Gene ID
363333
Gene Name
chromatin assembly factor 1, subunit A (p150)
Gene Aliases
-
Chromosome Location
Chr.9: 11000869 - 11027625 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_343671.6 XP_343672.2 13 - 14 2782 70

View Full Product Details