The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108895.1 | NP_001102365.1 | 6 - 7 | 833 | 67 |
XM_006255283.2 | XP_006255345.1 | 5 - 6 | 967 | 67 |