Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Egr1
Entrez Gene ID
24330
Gene Name
early growth response 1
Gene Aliases
Krox-24, NGFI-A, Ngf1, Ngfi, zif-268
Chromosome Location
Chr.18: 27657903 - 27660101 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012551.2 NP_036683.1 1 - 2 219 64
GenBank mRNA AY551092.1 - 1 - 2 219 64
M18416.1 - - 571 64

View Full Product Details