The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012558.3 | NP_036690.2 | 5 - 6 | 920 | 77 |
GenBank mRNA | BC078894.1 | - | 5 - 6 | 902 | 77 |
BC078895.1 | - | 5 - 6 | 920 | 77 | |
J04112.1 | - | 5 - 6 | 716 | 77 | |
M86240.1 | - | 5 - 6 | 806 | 77 |