The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012835.1 | NP_036967.1 | 1 - 2 | 157 | 149 |
GenBank mRNA | U51919.1 | - | 1 - 2 | 157 | 149 |