The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012987.1 | NP_037119.1 | 2 - 3 | 1040 | 78 |
XM_006232600.2 | XP_006232662.1 | 2 - 3 | 1358 | 78 | |
GenBank mRNA | AF538924.1 | - | 2 - 3 | 913 | 78 |
M34384.1 | - | 2 - 3 | 1040 | 78 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006232557.2 | XP_006232619.1 | 2 - 3 | 1359 | 78 |