Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sbp
Entrez Gene ID
25540
Gene Name
spermine binding protein
Gene Aliases
-
Chromosome Location
Chr.10: 13289514 - 13309332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767547.1 XP_008765769.1 1 - 2 225 66
GenBank mRNA J02675.1 - 1 - 2 249 66

View Full Product Details