The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013031.1 | NP_037163.1 | 10 - 11 | 1093 | 54 |
XM_006231664.2 | XP_006231726.1 | 10 - 11 | 1934 | 54 | |
XM_008760515.1 | XP_008758737.1 | 9 - 10 | 1293 | 54 | |
XM_008760516.1 | XP_008758738.1 | 10 - 11 | 1939 | 54 | |
XM_008760517.1 | XP_008758739.1 | 10 - 11 | 1934 | 54 | |
XM_008760518.1 | XP_008758740.1 | 9 - 10 | 1293 | 54 | |
XM_008760519.1 | XP_008758741.1 | 8 - 9 | 1200 | 54 | |
XM_008760520.1 | XP_008758742.1 | 7 - 8 | 1116 | 54 | |
GenBank mRNA | L00603.1 | - | 10 - 11 | 1093 | 54 |
M97381.1 | - | 10 - 11 | 1064 | 54 |