Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sycp3
Entrez Gene ID
25561
Gene Name
synaptonemal complex protein 3
Gene Aliases
RNASCP3
Chromosome Location
Chr.7: 29029887 - 29044344 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013041.1 NP_037173.1 3 - 4 316 149
GenBank mRNA X75785.1 - 3 - 4 316 149

View Full Product Details