Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mx2
Entrez Gene ID
286918
Gene Name
myxovirus (influenza virus) resistance 2
Gene Aliases
Mx3
Chromosome Location
Chr.11: 38035308 - 38062685 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134350.2 NP_599177.2 8 - 9 1123 86
XM_006248150.2 XP_006248212.1 10 - 11 1267 86
XM_008768568.1 XP_008766790.1 10 - 11 1201 86
GenBank mRNA BC099808.1 - 8 - 9 1123 86
X52712.1 - 8 - 9 1198 86
X52713.1 - 7 - 8 1003 86

View Full Product Details