The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017090.2 | NP_058786.2 | 6 - 7 | 1699 | 77 |
XM_006232518.2 | XP_006232580.1 | 7 - 8 | 2291 | 77 | |
XM_008761107.1 | XP_008759329.1 | 5 - 6 | 2322 | 77 | |
XM_008761108.1 | XP_008759330.1 | 6 - 7 | 2065 | 77 | |
GenBank mRNA | AB096020.1 | - | 5 - 6 | 1562 | 77 |
BC085746.1 | - | 6 - 7 | 1699 | 77 | |
M57405.1 | - | 6 - 7 | 2018 | 77 | |
U60835.1 | - | 6 - 7 | 2018 | 77 |