The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017103.2 | NP_058799.2 | 1 - 2 | 136 | 129 |
GenBank mRNA | BC081815.1 | - | 1 - 2 | 136 | 129 |
L46816.1 | - | - | 151 | 129 |