The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017206.1 | NP_058902.1 | 14 - 15 | 1852 | 140 |
XM_006236903.2 | XP_006236965.1 | 13 - 14 | 1961 | 140 | |
XM_008763123.1 | XP_008761345.1 | 15 - 16 | 2122 | 140 | |
GenBank mRNA | M96601.1 | - | 14 - 15 | 1852 | 140 |