The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017227.1 | NP_058923.1 | 1 - 2 | 112 | 65 |
XM_006239145.2 | XP_006239207.1 | 1 - 2 | 133 | 65 | |
GenBank mRNA | AB008803.1 | - | 1 - 2 | 112 | 65 |
AB010999.1 | - | 1 - 2 | 112 | 65 |